4-Bromo-o-toluonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-methylbenzonitrile

A173963-100G A173963-10G A173963-500G A173963-25G A173963-5G
103845-194EA 10 USD
103845-194 76769-310 76769-312 76769-314 77082-482 77082-484
4-Bromo-o-toluonitrile 98%
4-Bromo-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 259 °C (760 mm Hg)
Melting Pt: 65…69 °C
Flash Pt: 110.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00041461
CAS Number: 67832-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR