4-Bromo-o-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-methylbenzoic acid

A205937-100G A205937-10G A205937-1G A205937-500G A205937-25G A205937-5G
103848-342EA 7 USD
103848-342 76814-436 76814-438 76814-440 76814-442 77086-572 77086-574
4-Bromo-o-toluic acid 98%
4-Bromo-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 310.1 °C (760 mm Hg)
Melting Pt: 180…184 °C
Flash Pt: 141.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00040905
CAS Number: 68837-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR