4-Bromo-2-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A178549-1G A178549-5G A178549-100G A178549-10G A178549-25G
103807-350EA 6 USD
103807-350 103845-686 77083-116 77083-118 77287-078 77287-080 77301-562
4-Bromo-2-methylbenzaldehyde 98%
4-Bromo-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Boiling Pt: 264.6 °C (760 mm Hg)
Flash Pt: 97.627 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07787171
CAS Number: 24078-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR