4-Bromo-2-methyl-2H-1,2,3-triazole 95%

Supplier: Ambeed
Warning

A136090-100MG A136090-250MG
103802-272EA 33 USD
103802-272 76983-990 76983-992
4-Bromo-2-methyl-2H-1,2,3-triazole 95%
4-Bromo-2-methyl-2H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₃H₄BrN₃
Boiling Pt: 227.3±32.0 °C (760 mm Hg)
Melting Pt: 22 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD23378485
CAS Number: 16681-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR