5-Bromo-2-nitrotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-methylnitrobenzene, 4-Bromo-2-methyl-1-nitrobenzene

A221088-1G A221088-25G A221088-100G
103818-890EA 6 USD
103818-890 103849-770 77088-364 77088-366 77288-224
5-Bromo-2-nitrotoluene 97%
5-Bromo-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
Boiling Pt: 265.4 °C (760 mm Hg)
Melting Pt: 53…55 °C
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00137824
CAS Number: 52414-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR