4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine 98%
Supplier: Ambeed
|
Warning
|
A135658-100MG
A135658-250MG
103802-196EA
59
USD
103802-196
76782-644
76782-646
4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine 98%
4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.0586 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD15529172 CAS Number: 1014613-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...