4-Bromo-2-methyl-1H-indole 98%

Supplier: Ambeed
Warning

A263501-100MG A263501-250MG A263501-1G
103812-624EA 100 USD
103812-624 103852-512 76977-026 76977-028 77092-148
4-Bromo-2-methyl-1H-indole 98%
4-Bromo-2-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈BrN
Flash Pt: 151.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18380829
CAS Number: 6127-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR