4-Bromo-2-methoxypyridine 98%
Supplier: Ambeed
|
Warning
|
A131995-100G
A131995-10G
A131995-1G
A131995-25G
A131995-5G
103801-674EA
23
USD
103801-674
77285-352
77285-354
77314-212
77314-214
77314-216
4-Bromo-2-methoxypyridine 98%
4-Bromo-2-methoxypyridine
Precaución: for Research Use Only
Formula:
C₆H₆BrNO Boiling Pt: 202.3 °C (760 mm Hg) Flash Pt: 76.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08277268 CAS Number: 100367-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...