4-Bromo-2-methoxyphenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromoguaiacol, 5-Brom-2-hydroxyanisole

A119218-100G A119218-10G A119218-25G A119218-1G A119218-5G
103799-184EA 5 USD
103799-184 77284-738 77284-740 77284-742 77311-856 77311-858
4-Bromo-2-methoxyphenol 98%
4-Bromo-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
MW: 203.04 g/mol
Boiling Pt: 250.1 °C (760 mm Hg)
Melting Pt: 34…37 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051937
CAS Number: 7368-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR