4-Bromo-2-methoxybenzyl alcohol 97%

Supplier: Ambeed
Warning

A471029-10G A471029-250MG
103828-026EA 13 USD
103828-026 103839-100 76979-672 76979-674
4-Bromo-2-methoxybenzyl alcohol 97%
4-Bromo-2-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 290.6 °C (760 mm Hg)
Melting Pt: 66…70 °C
Flash Pt: 129.5 °C
Storage Temperature: Ambient
MDL Number: MFCD07369762
CAS Number: 17102-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR