4-Bromo-o-anisaldehyde 98%

Supplier: Ambeed
Warning

A217710-1G A217710-25G A217710-5G A217710-100G A217710-10G A217710-250MG
103818-638EA 6 USD
103818-638 103849-368 77087-994 77087-996 77087-998 77288-122 77288-124 77288-126
4-Bromo-o-anisaldehyde 98%
4-Bromo-o-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
Boiling Pt: 288.1 °C (760 mm Hg)
Melting Pt: 67…71 °C
Flash Pt: 128.042 °C
Storage Temperature: Ambient
MDL Number: MFCD08276823
CAS Number: 43192-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR