4-Bromo-2-methoxy-3-methylpyridine 98%

Supplier: Ambeed
Warning

A214597-250MG A214597-1G
103818-418EA 299 USD
103818-418 103849-122 76814-676 77087-652
4-Bromo-2-methoxy-3-methylpyridine 98%
4-Bromo-2-methoxy-3-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16610111
CAS Number: 112197-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR