5-Bromo-2-methylanisole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-methoxy-1-methylbenzene

A156132-1G A156132-5G A156132-10G A156132-100G
103804-830EA 5 USD
103804-830 103843-034 77318-776 77318-778 77592-918 77671-202
5-Bromo-2-methylanisole 97%
5-Bromo-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797972
CAS Number: 67868-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR