4-Bromo-2-iodophenol 97%

Supplier: Ambeed
Warning

A106990-100G A106990-10G A106990-250MG A106990-25G A106990-1G A106990-5G
103797-106EA 7 USD
103797-106 103822-864 77283-524 77283-526 77283-528 77283-530 77309-734 77309-736
4-Bromo-2-iodophenol 97%
4-Bromo-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
Boiling Pt: 243 °C (760 mm Hg)
Melting Pt: 71 °C
Flash Pt: 100.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09029644
CAS Number: 207115-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR