4-Bromo-2-iodobenzaldehyde 97%

Supplier: Ambeed
Warning

A139500-250MG A139500-1G
103802-748EA 29 USD
103802-748 76984-132 77315-842
4-Bromo-2-iodobenzaldehyde 97%
4-Bromo-2-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrIO
MW: 310.915 g/mol
Boiling Pt: 642.7 °C (760 mm Hg)
Flash Pt: 342.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18391619
CAS Number: 1261470-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR