4-Bromo-2-iodoaniline 97%

Supplier: Ambeed
Danger

A208029-1G A208029-25G A208029-5G A208029-10G A208029-250MG
103817-848EA 8 USD
103817-848 103848-534 77086-834 77086-836 77086-838 77287-874 77287-876
4-Bromo-2-iodoaniline 97%
4-Bromo-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅BrIN
Melting Pt: 69…72 °C
Flash Pt: 134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093764
CAS Number: 66416-72-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR