4-Bromo-2-iodotoluene 98%
Supplier: Ambeed
|
Warning
|
A512314-5G
A512314-100G
A512314-10G
A512314-25G
A512314-500G
A512314-1G
103828-408EA
5
USD
103828-408
103839-508
76843-494
77034-604
77034-606
77034-608
77034-610
77307-662
4-Bromo-2-iodotoluene 98%
4-Bromo-2-iodotoluene
Precaución: for Research Use Only
Formula:
C₇H₆BrI Boiling Pt: 268.4 °C (760 mm Hg) Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780671 CAS Number: 260558-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...