4-Bromosalicylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-2-hydroxybenzoic acid
A321413-1G
A321413-25G
A321413-5G
A321413-100G
A321413-10G
A321413-500G
103825-272EA
9
USD
103825-272
103836-206
76839-100
76839-102
76839-104
77141-806
77141-808
77141-810
4-Bromosalicylic acid 98%
4-Bromosalicylic acid
Precaución: for Research Use Only
Formula:
C₇H₅BrO₃ MW: 217.02 g/mol Boiling Pt: 330.2 °C (760 mm Hg) Melting Pt: 216…217 °C Flash Pt: 153.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797970 CAS Number: 1666-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...