4-Bromo-2-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A231337-10G
A231337-5G
A231337-1G
A231337-25G
A231337-100G
103810-772EA
6
USD
103810-772
103850-520
76987-394
77089-408
77303-908
77303-910
77550-498
4-Bromo-2-fluoropyridine 98%
4-Bromo-2-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₃BrFN Boiling Pt: 195.9 °C (760 mm Hg) Flash Pt: 71.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD04112504 CAS Number: 128071-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...