4-Bromo-2-fluorophenol 98%

Supplier: Ambeed
Warning

A419367-25G A419367-100G A419367-10G A419367-500G A419367-5G
103838-412EA 5 USD
103838-412 76842-026 77291-380 77291-382 77291-384 77307-200
4-Bromo-2-fluorophenol 98%
4-Bromo-2-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrFO
Boiling Pt: 206.1 °C (760 mm Hg)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00011722
CAS Number: 2105-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR