4-Bromo-2-fluoro-N-methylbenzamide 98%
Supplier: Ambeed
|
Warning
|
A347498-1G
A347498-5G
103825-968EA
5
USD
103825-968
103836-954
76840-032
76840-034
4-Bromo-2-fluoro-N-methylbenzamide 98%
4-Bromo-2-fluoro-N-methylbenzamide
Precaución: for Research Use Only
Formula:
C₈H₇BrFNO Boiling Pt: 284.8 °C (760 mm Hg) Flash Pt: 126.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD09878362 CAS Number: 749927-69-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...