1-Bromo-3-fluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-fluoronitrobenzene

A402858-25G A402858-5G A402858-100G A402858-10G A402858-1G A402858-500G
103838-120EA 6 USD
103838-120 76841-590 76841-592 77291-152 77291-154 77291-156 77291-158
1-Bromo-3-fluoro-4-nitrobenzene 98%
1-Bromo-3-fluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Boiling Pt: 263.2 °C (760 mm Hg)
Melting Pt: 84…88 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD01930221
CAS Number: 321-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR