4-Bromo-2-fluoro-3-formylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-fluoronicotinaldehyde

A328064-10G A328064-250MG A328064-1G A328064-5G
103825-492EA 22 USD
103825-492 103836-446 76978-200 76978-202 77306-372 77306-374
4-Bromo-2-fluoro-3-formylpyridine 97%
4-Bromo-2-fluoro-3-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₃BrFNO
MW: 204 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09870050
CAS Number: 128071-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR