4-Bromo-2-fluorobenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A407052-25G
A407052-5G
A407052-1000G
A407052-100G
A407052-10G
A407052-500G
103838-186EA
5
USD
103838-186
76841-674
76841-676
77115-776
77115-778
77115-780
77115-782
4-Bromo-2-fluorobenzonitrile 98%
4-Bromo-2-fluorobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃BrFN Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143264 CAS Number: 105942-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...