4-Bromo-2-fluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A165322-1000G
A165322-100G
A165322-10G
A165322-500G
A165322-5G
A165322-25G
103844-212EA
6
USD
103844-212
77273-512
77273-514
77273-516
77273-518
77300-772
77320-414
4-Bromo-2-fluorobenzoic acid 98%
4-Bromo-2-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrFO₂ Melting Pt: 211…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143263 CAS Number: 112704-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...