4-Bromo-2-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A119816-100G A119816-10G A119816-25G A119816-500G A119816-5G
76766-406EA 59.42 USD
76766-406 76766-408 76766-410 76766-412 77311-954
4-Bromo-2-fluorobenzaldehyde 98%
4-Bromo-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO
MW: 203.01 g/mol
Boiling Pt: 241.4 °C (760 mm Hg)
Melting Pt: 59…64 °C
Flash Pt: 99.801 °C
Storage Temperature: Ambient
MDL Number: MFCD00143261
CAS Number: 57848-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR