4-Bromo-2-fluoro-6-nitrotoluene 98%

Supplier: Ambeed
Warning

A329275-100MG A329275-10G A329275-250MG A329275-1G A329275-25G A329275-5G A329275-100G
103825-522EA 12 USD
103825-522 103836-476 76817-132 76817-134 76817-136 76839-442 76839-444 76839-446 77672-692
4-Bromo-2-fluoro-6-nitrotoluene 98%
4-Bromo-2-fluoro-6-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
Boiling Pt: 261.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03094190
CAS Number: 502496-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR