4-Bromo-2-fluoro-6-methoxybenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A164040-100MG
A164040-250MG
A164040-1G
103805-802EA
95
USD
103805-802
103844-068
77011-342
77011-344
77320-198
4-Bromo-2-fluoro-6-methoxybenzoic acid 97%
4-Bromo-2-fluoro-6-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD22570393 CAS Number: 1472104-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...