4-Bromo-2-fluoro-6-methoxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A166880-10G
A166880-250MG
A166880-1G
A166880-5G
103806-108EA
59
USD
103806-108
103844-370
77011-440
77011-442
77320-652
77320-654
4-Bromo-2-fluoro-6-methoxybenzaldehyde 97%
4-Bromo-2-fluoro-6-methoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ MW: 233.034 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22543644 CAS Number: 856767-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...