4-Bromo-2-fluoro-6-hydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A142114-10G A142114-250MG A142114-1G
103803-102EA 149 USD
103803-102 77010-392 77010-394 77316-316
4-Bromo-2-fluoro-6-hydroxybenzaldehyde 98%
4-Bromo-2-fluoro-6-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD23705560
CAS Number: 1427438-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR