4-Bromo-2-fluoro-5-methylbenzoic acid 98%

Supplier: Ambeed
Warning

A263091-1G A263091-25G A263091-5G A263091-100G A263091-100MG A263091-10G A263091-250MG
103812-606EA 8 USD
103812-606 103852-502 77092-134 77092-136 77092-138 77289-256 77289-258 77289-260 77289-262
4-Bromo-2-fluoro-5-methylbenzoic acid 98%
4-Bromo-2-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.0300 g/mol
Boiling Pt: 302.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08458245
CAS Number: 415965-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR