4-Bromo-2-fluoro-3-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A215661-100MG
A215661-10G
A215661-250MG
A215661-1G
103818-506EA
30
USD
103818-506
103849-214
77261-188
77261-190
77261-192
77303-416
4-Bromo-2-fluoro-3-methylpyridine 98%
4-Bromo-2-fluoro-3-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrFN Boiling Pt: 220.2 °C (760 mm Hg) Flash Pt: 87 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09839278 CAS Number: 128071-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...