4-Bromo-2-fluoro-3-methylbenzonitrile 97%

Supplier: Ambeed
Warning

A108513-100MG A108513-250MG A108513-1G
103797-360EA 334 USD
103797-360 103823-140 77160-598 77160-600 77309-986
4-Bromo-2-fluoro-3-methylbenzonitrile 97%
4-Bromo-2-fluoro-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD23707391
CAS Number: 1114546-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR