4-Bromo-2-fluoro-3-methylbenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A149126-100MG
A149126-250MG
A149126-1G
A149126-5G
103804-004EA
40
USD
103804-004
103842-128
77162-696
77162-698
77299-706
77299-708
4-Bromo-2-fluoro-3-methylbenzoic acid 95%
4-Bromo-2-fluoro-3-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ MW: 233.034 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22052783 CAS Number: 194804-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...