1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A122429-100G A122429-10G A122429-1G A122429-25G A122429-500G A122429-5G
77026-506EA 144 USD
77026-506 77026-508 77026-510 77026-512 77026-514 77312-360
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene 98%
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄O
Boiling Pt: 175.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01861132
CAS Number: 105529-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR