4-Bromo-2-cyclopropylpyridine 97%

Supplier: Ambeed
Warning

A342529-1G A342529-250MG
103825-864EA 263 USD
103825-864 76839-882 76989-464
4-Bromo-2-cyclopropylpyridine 97%
4-Bromo-2-cyclopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₈BrN
MW: 198.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11223210
CAS Number: 1086381-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR