4-Bromo-2-chloropyrimidine 95%

Supplier: Ambeed
Warning

A243628-250MG A243628-25G
103811-528EA 65 USD
103811-528 103851-350 76976-528 77672-178
4-Bromo-2-chloropyrimidine 95%
4-Bromo-2-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂
Boiling Pt: 291.552 °C (760 mm Hg)
Flash Pt: 130.126 °C
Storage Temperature: Freezer
MDL Number: MFCD08272196
CAS Number: 885702-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR