4-Bromo-2-chloropyridine 98%

Supplier: Ambeed
Warning

A182631-25G A182631-5G A182631-100G A182631-10G A182631-1G A182631-500G
103846-110EA 8 USD
103846-110 77083-658 77083-660 77274-142 77274-144 77274-146 77615-546
4-Bromo-2-chloropyridine 98%
4-Bromo-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03840756
CAS Number: 73583-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR