4-Bromo-2-chloronicotinonitrile 95%
Supplier: Ambeed
|
Warning
|
A223764-10G
A223764-250MG
A223764-1G
103819-060EA
89
USD
103819-060
103849-942
77112-800
77112-802
77303-672
4-Bromo-2-chloronicotinonitrile 95%
4-Bromo-2-chloronicotinonitrile
Precaución: for Research Use Only
Formula:
C₆H₂BrClN₂ MW: 217.45 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD27937928 CAS Number: 1807017-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...