4-Bromo-2-chlorobenzotrifluoride 98%

Supplier: Ambeed
Warning

A533989-25G A533989-100G A533989-500G
103839-702EA 6 USD
103839-702 76843-716 77116-746 77116-748
4-Bromo-2-chlorobenzotrifluoride 98%
4-Bromo-2-chlorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 224.1 °C (760 mm Hg)
Flash Pt: 89.3 °C
Storage Temperature: Ambient
MDL Number: MFCD08459292
CAS Number: 467435-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR