4-Bromo-2-chlorobenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A145270-250MG A145270-5G
103803-512EA 44 USD
103803-512 103841-594 76997-022 77299-442
4-Bromo-2-chlorobenzo[d]thiazole 97%
4-Bromo-2-chlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₃BrClNS
MW: 248.5300 g/mol
Boiling Pt: 309.2 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD04971817
CAS Number: 182344-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR