4-Bromo-6-chloro-o-cresol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-chloro-6-methylphenol

A434122-25G A434122-5G A434122-10G A434122-1G A434122-100G
103827-570EA 5 USD
103827-570 103838-638 76842-352 76842-354 77291-562 77307-274 77748-878
4-Bromo-6-chloro-o-cresol 98%
4-Bromo-6-chloro-o-cresol

Precaución: for Research Use Only

Formula: C₇H₆BrClO
Boiling Pt: 254 °C (760 mm Hg)
Melting Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00002167
CAS Number: 7530-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR