4-Bromo-2-fluoro-6-chlorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-Chloro-6-fluorobenzoic acid

A122558-10G A122558-250MG A122558-25G A122558-1G A122558-5G A122558-100G
103800-240EA 17 USD
103800-240 77094-546 77094-548 77094-550 77312-370 77312-372 77901-959
4-Bromo-2-fluoro-6-chlorobenzoic acid 97%
4-Bromo-2-fluoro-6-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
Boiling Pt: 303.0±42.0 °C (760 mm Hg)
Flash Pt: 137 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18917163
CAS Number: 1321613-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR