4-Bromo-2-chloro-5-nitropyridine 95%

Supplier: Ambeed
Warning

A728331-100MG A728331-10G A728331-250MG A728331-1G A728331-5G
103840-774EA 548 USD
103840-774 77118-078 77118-080 77118-082 77308-218 77308-220
4-Bromo-2-chloro-5-nitropyridine 95%
4-Bromo-2-chloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD17014986
CAS Number: 1261767-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR