4-Bromo-2-chloro-5-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
A120940-100MG
A120940-10G
A120940-250MG
A120940-1G
A120940-5G
103800-012EA
30
USD
103800-012
76995-830
76995-832
76995-834
77312-122
77312-124
4-Bromo-2-chloro-5-methylpyridine 97%
4-Bromo-2-chloro-5-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrClN MW: 206.46 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11100650 CAS Number: 867279-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...