4-Bromo-2-chloro-5-methoxypyridine 95%

Supplier: Ambeed
Warning

A230972-100MG A230972-250MG
103810-740EA 119 USD
103810-740 103850-486 76874-242 76874-244
4-Bromo-2-chloro-5-methoxypyridine 95%
4-Bromo-2-chloro-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
Boiling Pt: 267.3±35.0 °C (760 mm Hg)
Flash Pt: 115.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09972186
CAS Number: 1020253-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR