4-Bromo-2-chloro-1-nitrobenzene 95%

Supplier: Ambeed
Warning

A789979-1G A789979-100MG A789979-250MG
103841-012EA 189 USD
103841-012 76845-266 76993-224 76993-226
4-Bromo-2-chloro-1-nitrobenzene 95%
4-Bromo-2-chloro-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
Boiling Pt: 286.1±20.0 °C (760 mm Hg)
Melting Pt: 42.4 °C
Flash Pt: 126.8 °C
Storage Temperature: Ambient
MDL Number: MFCD11848538
CAS Number: 89465-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR