4-Bromo-2-benzofuran-1[3H]-one 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromophthalide

A366898-100MG A366898-250MG
103826-414EA 46 USD
103826-414 103837-428 77290-860 77290-862
4-Bromo-2-benzofuran-1[3H]-one 97%
4-Bromo-2-benzofuran-1[3H]-one

Precaución: for Research Use Only

Formula: C₈H₅BrO₂
Boiling Pt: 378.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06412577
CAS Number: 102308-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR