4-Bromo-2,6-di-tert-butylphenol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,5-di(dimethylethyl)-4-hydroxybenzene

A105089-100G A105089-25G A105089-500G
103822-564EA 9 USD
103822-564 76994-808 76994-810 76994-812
4-Bromo-2,6-di-tert-butylphenol 98%
4-Bromo-2,6-di-tert-butylphenol

Precaución: for Research Use Only

Formula: C₁₄H₂₁BrO
MW: 285.22 g/mol
Melting Pt: 78…83 °C
Flash Pt: 119.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00051598
CAS Number: 1139-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR