4-Bromo-2,6-dimethylpyridine 98%
Supplier: Ambeed
|
Warning
|
A267528-10G
A267528-1G
A267528-25G
A267528-5G
A267528-100G
103812-858EA
9
USD
103812-858
103852-758
76988-268
77092-504
77092-506
77092-508
77550-672
4-Bromo-2,6-dimethylpyridine 98%
4-Bromo-2,6-dimethylpyridine
Precaución: for Research Use Only
Formula:
C₇H₈BrN MW: 186.0491 g/mol Melting Pt: 28…32 °C Flash Pt: 79.2 °C Storage Temperature: Freezer |
MDL Number:
MFCD00671475 CAS Number: 5093-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...